Learn More
Agilent Technologies BIO-INERT CAP UHP-FF FITTING
Catalog No. NC2168138
Encompass
UHP-FF fitting. for use with Stainless Steel-coated PEEK capillaries, bio-inert
This item is not returnable. View return policy
Catalog No. NC2168138 Supplier Agilent Technologies Supplier No. 50675695
UHP-FF fitting. for use with Stainless Steel-coated PEEK capillaries, bio-inert