Learn More
Agilent Technologies BIO-INERT STANDARD FLOW CELL
Catalog No. NC1492965
Encompass
nfinityLab bio-inert standard flow cell, with RFID tag, 10 mm, 13 L, 120 bar,
Catalog No. NC1492965 Supplier Agilent Technologies Supplier No. G561560022
nfinityLab bio-inert standard flow cell, with RFID tag, 10 mm, 13 L, 120 bar,