Learn More
RESONON INC Bio-LIF system combines Laser-Induced Fluorescence and Hyperspectral Imaging to yield unparalleled spectral resolution of emission data and insight into biological samples.
SDP
Catalog No. NC2953205
Encompass_Preferred
Resonon's Bio-LIF system used Hyperspectral Imaging to provide detailed spectral information of Laser Induced Fluorescent emissions. Features 335 spectral channels for each pixel, enabling straightforward discrimination of shifted or overlapping emissions. Automated scan routine with built-in spectral calibration and auto-exposure. Tray carrier accepts 90mm Ø dishes or standard 96-well microplates. Scan time for a microplate is typically around 30 seconds.
Catalog No. NC2953205 Supplier RESONON INC Supplier No. RES0003
Add to Cart
Add to Cart
Request a Quote
Resonon's Bio-LIF system used Hyperspectral Imaging to provide detailed spectral information of Laser Induced Fluorescent emissions. Features 335 spectral channels for each pixel, enabling straightforward discrimination of shifted or overlapping emissions. Automated scan routine with built-in spectral calibration and auto-exposure. Tray carrier accepts 90mm Ø dishes or standard 96-well microplates. Scan time for a microplate is typically around 30 seconds.