Learn More
Optimize Technologies OPTI-SOLV™ Direct Connect Biocompatible Nano Filter, 0.5µm, 5pk
SDP
Supplier:  Optimize Technologies 100403939
Encompass_Preferred
The OPTI-SOLV™ Nano Filter cuts the internal volume down even further. With less than 50nL of internal volume, the OPTI-SOLV™ Nano Filter is ideal for mass spectrometry to protect electrospray and nanospray tips from clogging with debris. The Nano Filter is designed to be used in today’s low volume, high sensitivity chromatographic applications as a last line of defense against debris. As with all Optimize column protection products, the filter features a zero-dead-volume connection utilizing patented floating stem (auto-adjusting) technology. The efficient design requires no holder and threads directly into any 10-32 port. It is available in 5 packs and recommended for volume-critical applications. For use up to 6,000psi.
Catalog No. 50-200-6158