Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio Plas™ Foam Biopsy Pads

Catalog No. 22228379
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-228379 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-228379 Supplier Bio Plas™ Supplier No. 6130

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For histology specimen protection

  • Fabricated from cellular polyester plastic foam
  • Air pockets throughout pad allow for adequate fluid movement around tissue while securely sandwiched between two pads
  • Assure the orientation of small biopsy tissue
  • White
  • For use with 1 × 1 1/4 in. cassettes

Specifications

Material Cellular Polyester Urethane Foam
Color White

Latex free

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.