Learn More
Bio Rad Laboratories BIO-PLEX CELL LYSIS KIT
Catalog No. 509010008
Encompass
Kit, Cell Lysis; Bio-Rad; Bio-Plex; Sample preparation kit; Includes cell lysis buffer, cell wash buffer, factor 1 and factor 2; 1 x 96-well
Catalog No. 50-901-0008 Supplier Bio Rad Laboratories Supplier No. 171304011
Kit, Cell Lysis; Bio-Rad; Bio-Plex; Sample preparation kit; Includes cell lysis buffer, cell wash buffer, factor 1 and factor 2; 1 x 96-well