Learn More
Enzo Life Sciences BIO-PROBE Cytomegalovirus probe (2µg)
SDP
Catalog No. 502012506
Encompass_Preferred
The Cytomegalovirus (CMV) BIO-PROBE labeled probe is prepared by nick translation of cloned fragments of the CMV Towne strain. The fragments include a total of 30-31 kb of DNA, approximately 20% of the CMV genome. The probe is specific for cytomegalovirus DNA. It does not hybridize to DNAs of other herpesviruses (i.e., herpes simplex virus or Epstein-Barr virus) but it has been found to hybridize to the DNA of CMV clinical isolates. Fragment size range: 100-1000 base pairs (as estimated by agarose gel electrophoresis). Concentration: 20µg/ml. Formulation: Liquid. In 10mM TRIS HCl, pH 7.5, containing 1mM EDTA. Long Term Storage: -20°C.
Catalog No. 50-201-2506 Supplier Enzo Life Sciences Supplier No. ENZ40835
The Cytomegalovirus (CMV) BIO-PROBE labeled probe is prepared by nick translation of cloned fragments of the CMV Towne strain. The fragments include a total of 30-31 kb of DNA, approximately 20% of the CMV genome. The probe is specific for cytomegalovirus DNA. It does not hybridize to DNAs of other herpesviruses (i.e., herpes simplex virus or Epstein-Barr virus) but it has been found to hybridize to the DNA of CMV clinical isolates. Fragment size range: 100-1000 base pairs (as estimated by agarose gel electrophoresis). Concentration: 20µg/ml. Formulation: Liquid. In 10mM TRIS HCl, pH 7.5, containing 1mM EDTA. Long Term Storage: -20°C.