Learn More
Bellco Glass, Inc. Ideal for small working volumes with ph and do probe. 32mm low sidearms for insertion. Size: 1l Center cap size: 100mm 32mm compression fitting probe & port assemblies sold separately
SDP
Catalog No. 501945127
Encompass_Preferred
Ideal for small working volumes with ph and do probe. 32mm low sidearms for insertion. Size: 1l Center cap size: 100mm 32mm compression fitting probe & port assemblies sold separately
Catalog No. 50-194-5127 Supplier Bellco Glass, Inc. Supplier No. 196597001
Ideal for small working volumes with ph and do probe. 32mm low sidearms for insertion. Size: 1l Center cap size: 100mm 32mm compression fitting probe & port assemblies sold separately