Learn More
Bellco Glass, Inc. Bio-Probe Complete,500mL
SDP
Catalog No. 501945129
Encompass_Preferred
Ideal for small working volumes with ph and do probe. 32mm low sidearms for insertion.. MADE IN THE USA Size: 500ml Center cap size: 100mm 32mm compression fitting probe & port assemblies sold separately
Catalog No. 50-194-5129 Supplier Bellco Glass, Inc. Supplier No. 196597005
Ideal for small working volumes with ph and do probe. 32mm low sidearms for insertion.. MADE IN THE USA Size: 500ml Center cap size: 100mm 32mm compression fitting probe & port assemblies sold separately