Learn More
Enzo Life Sciences BIO-PROBE Epstein-Barr virus (EBV) probe (2µg)
SDP
Supplier:  Enzo Life Sciences ENZ40836
Encompass_Preferred
The BIO-PROBE Epstein-Barr virus (EBV) labeled probe is prepared by nick translation of the 2.68 Bam HI "V" region (the internal repeat) of the EBV genome. The Bam HI clone is contained in pUC 18 vector. The probe does not cross hybridize with the other herpes virus DNAs, herpes simplex virus DNA or cytomegalovirus DNA. Concentration: 20 µg/mL. Formulation: Liquid. In 10mM TRIS HCl, pH 7.5, containing 1mM EDTA. Long Term Storage: -20°C.
Catalog No. 50-201-2507