Learn More
Enzo Life Sciences BIO-PROBE Lambda probe (2µg)
SDP
Supplier:  Enzo Life Sciences ENZ40840
Encompass_Preferred
The Lambda BIO-PROBE labeled probe is prepared by nick translation of the lambda bacteriophage strain c1857ind1 Sam7. Fragment size range: 100-1000 base pairs (as estimated by agarose gel electrophoresis). Concentration: 20µg/ml. Formulation: Liquid. In 10mM TRIS-HCl, pH 7.5, containing 1mM EDTA. Long Term Storage: -20°C.
Catalog No. 50-201-2509