Learn More
Bellco Glass, Inc. Bio-Probe Spinner,1L
SDP
Catalog No. 501945132
Encompass_Preferred
•Material: Type 1 Class A Borosilicate Glass . MADE IN THE USA•Dimensions: 136mm OD x 240mm Overall Height•Two 45mm Side arms angled 60° off center•Two 32mm Side arms angled 35° off center•One 100mm Screw Thread Center Neck•Indented bottom•Graduations: 200mL to 1000mL, every 100mL – White Ink (Approx Volumes)•Supply with: Two 45mm PP Blue Screw Caps, two PP Blue Pouring Rings, two 32mm blue screw caps, and two o-rings
Catalog No. 50-194-5132 Supplier Bellco Glass, Inc. Supplier No. 196597100
•Material: Type 1 Class A Borosilicate Glass . MADE IN THE USA•Dimensions: 136mm OD x 240mm Overall Height•Two 45mm Side arms angled 60° off center•Two 32mm Side arms angled 35° off center•One 100mm Screw Thread Center Neck•Indented bottom•Graduations: 200mL to 1000mL, every 100mL – White Ink (Approx Volumes)•Supply with: Two 45mm PP Blue Screw Caps, two PP Blue Pouring Rings, two 32mm blue screw caps, and two o-rings