Learn More
Bellco Glass, Inc. Bio-Probe Spinner,3L
SDP
Catalog No. 501945133
Encompass_Preferred
•Material: Type 1 Class A Borosilicate Glass. MADE IN THE USA•Dimensions: 168mm OD x 290mm Overall Height•Two 45mm Side arms angled 53° off center•Two 32mm Side arms angled 35° off center•One 100mm Screw Thread Center Neck•Indented bottom•Graduations: 500mL to 3000mL, every 500mL – White Ink (Approx Volumes)•Supply with: Two 45mm PP Blue Screw Caps, two PP Blue Pouring Rings, two 32mm blue screw caps, and two o-rings
Catalog No. 50-194-5133 Supplier Bellco Glass, Inc. Supplier No. 196597300
•Material: Type 1 Class A Borosilicate Glass. MADE IN THE USA•Dimensions: 168mm OD x 290mm Overall Height•Two 45mm Side arms angled 53° off center•Two 32mm Side arms angled 35° off center•One 100mm Screw Thread Center Neck•Indented bottom•Graduations: 500mL to 3000mL, every 500mL – White Ink (Approx Volumes)•Supply with: Two 45mm PP Blue Screw Caps, two PP Blue Pouring Rings, two 32mm blue screw caps, and two o-rings