Learn More
Electron Microscopy Sciences Bio-Pure™ Alcohol Wipes
SDP
Catalog No. 5034508
Encompass_Preferred
Proprietary durable fabric formulated to easily break down over time making it 100% biodegradable. The re-sealable and flexible packaging has a low carbon footprint. 70% IPA/30% H2O mixture exaporates completely, leaving no residue. Wipes won't scratch surfaces or optics. Clean lab surfaces, equipment, optical parts and cuvettes.
This item is not returnable. View return policy
Catalog No. 50-345-08 Supplier Electron Microscopy Sciences Supplier No. 6855214
Proprietary durable fabric formulated to easily break down over time making it 100% biodegradable. The re-sealable and flexible packaging has a low carbon footprint. 70% IPA/30% H2O mixture exaporates completely, leaving no residue. Wipes won't scratch surfaces or optics. Clean lab surfaces, equipment, optical parts and cuvettes.