Learn More
Sigma Aldrich Fine Chemicals Biosciences Bio-PureTM pipetting reser
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z679933100EA
Encompass
Biodegradable - Using a special additive to PET our reservoirs will safely degrade over timeTrough within a trough maximizes reagent recoveryPour-off spouts on all four corners and a wide base and reinforced sidewalls avoids spills and provides stabilitySterile
Catalog No. 50-290-1522