Learn More
Sigma Aldrich Fine Chemicals Biosciences Bio-PureTM pipetting reser
Catalog No. 502900354
Encompass
Biodegradable - Using a special additive to PET our reservoirs will safely degrade over timeTrough within a trough maximizes reagent recoveryPour-off spouts on all four corners and a wide base and reinforced sidewalls avoids spills and provides stabilitySterile
Catalog No. 50-290-0354 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z680354100EA
Biodegradable - Using a special additive to PET our reservoirs will safely degrade over timeTrough within a trough maximizes reagent recoveryPour-off spouts on all four corners and a wide base and reinforced sidewalls avoids spills and provides stabilitySterile