Learn More
Bio Rad Laboratories Bio-Rad Dual 48/48 Fast Reaction Module, Gradient Enabled, Independent Dual Well, For Thermal Cyclers, 1 ea
Catalog No. NC3929582
Encompass
This independent dual 48/48 fast reaction module is a swappable thermal cycling component designed to run two distinct PCR protocols simultaneously. Compatible with specific thermal cycler chassis, it enables high-throughput flexibility by allowing independent temperature control for up to 48 samples per side.
  • Supports two independent pcr protocols for up to 48 samples each
  • Includes gradient capabilities for optimization
  • Features a dual 48-well design for increased experimental efficiency
  • Compatible with selected thermal cycler systems
  • Accommodates 0.2 ml tubes or 48-well plates
  • For research use only
Catalog No. NC3929582 Supplier Bio Rad Laboratories Supplier No. 1840148
This independent dual 48/48 fast reaction module is a swappable thermal cycling component designed to run two distinct PCR protocols simultaneously. Compatible with specific thermal cycler chassis, it enables high-throughput flexibility by allowing independent temperature control for up to 48 samples per side.
  • Supports two independent pcr protocols for up to 48 samples each
  • Includes gradient capabilities for optimization
  • Features a dual 48-well design for increased experimental efficiency
  • Compatible with selected thermal cycler systems
  • Accommodates 0.2 ml tubes or 48-well plates
  • For research use only