Learn More
Bio Rad Laboratories BIO-RAD Econo-Column Chromatography Column, Glass, 1.0 × 5 cm, 4 mL
Supplier:  Bio Rad Laboratories 7371007
Encompass
These glass chromatography columns are designed for use with low-pressure (<1 bar) chromatography systems or gravity flow separations. Ideal for simple packing and procedure running, this column has a 4 mL volume and dimensions of 1.0 × 5 cm.
  • Porous polymer bed support at the bottom of the column retains fine particles.
  • Translucent polypropylene end fittings allow viewing of the entire column bed.
  • Autoclavable and sodium hydroxide (NaOH) compatible.
  • Includes one stopcock per column.
Catalog No. NC3996987