Learn More
Bio Rad Laboratories BIO-RAD PROTEIN ASSAY KIT I
Catalog No. 509012439
Encompass
Kit I; Protein Assay; Bio-Rad; Contains 450mL dye reagent concentrate and a bovine gamma-globulin standard; Sufficient for 440 standard assays or 2,200 microplate assays
Catalog No. 50-901-2439 Supplier Bio Rad Laboratories Supplier No. 5000001
Kit I; Protein Assay; Bio-Rad; Contains 450mL dye reagent concentrate and a bovine gamma-globulin standard; Sufficient for 440 standard assays or 2,200 microplate assays