Learn More
Bio Rad Laboratories BIO-RAD PROTIEN ASSAY KIT II
Catalog No. NC9312385
Encompass
Bio-Rad Protein Assay Kit II, Contains 450mL dye reagent concentrate, bovine serum albumin standard, based on method of Bradford, A colorimetric assay for measuring total protein concentration
Catalog No. NC9312385 Supplier Bio Rad Laboratories Supplier No. 5000002
Bio-Rad Protein Assay Kit II, Contains 450mL dye reagent concentrate, bovine serum albumin standard, based on method of Bradford, A colorimetric assay for measuring total protein concentration