Learn More
Bio Rad Laboratories Sub-Cell GT Gel Caster
Catalog No. NC1950336
Encompass
Use the Sub-Cell GT Gel Caster to cast agarose gels for the Sub-Cell GT System. Prepare your agarose gels conveniently ahead of time without tying up your Sub-Cell workflow.
Catalog No. NC1950336 Supplier Bio Rad Laboratories Supplier No. 1704412
Use the Sub-Cell GT Gel Caster to cast agarose gels for the Sub-Cell GT System. Prepare your agarose gels conveniently ahead of time without tying up your Sub-Cell workflow.