Learn More
Chemglass Life Sciences 7L Unjacketed Bio Reactor Vessel
SDP
Catalog No. 501294475
Encompass
7L Unjacketed Bio Reactor Vessel. Vessels are constructed of heavy wall tubing and are graduated in 200mL subdivisions with numbers every 1000mL. Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors. Applikon is a registered trademark of Applikon Biotechnology, Inc. Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.
Catalog No. 50-129-4475 Supplier Chemglass Life Sciences Supplier No. CLS138301
7L Unjacketed Bio Reactor Vessel. Vessels are constructed of heavy wall tubing and are graduated in 200mL subdivisions with numbers every 1000mL. Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors. Applikon is a registered trademark of Applikon Biotechnology, Inc. Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.