Learn More
Chemglass Life Sciences Bio Reactor Accessory Kit Complete, Platinum Cured Silicone Tubing
SDP
Supplier:  Chemglass Life Sciences CLS139001
Encompass
This pre fabricated, single use bio reactor kit consists of the 8 components listed.They are packed in an ISO Class 7 clean room, double bagged and ready for use.Kits are fabricated with full traceability and accompanying validation documentation, which meets USP Class VI requirements.These pre-assembled kits can be customized to any validated components currently specified on your bioreactor design.Features/Benefits: Single Use: Minimize Labor Costs.Eliminates Cleaning Validation.Reduces Set up Time and Costs.Pre-Fabricated Assemblies: Minimize Injuries associated with cutting in-house.Improved consistency.Offers Closed System: Gives the ability to assemble in Open Lab type Environment.How to Sterilize: Autoclave Max 15 psi @ 134 degreesC, 60 min.Gamma Radiation 25 kGy to 50 kGy.Applikon is a registered trademark of Applikon Biotechnology, Inc.Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.
Catalog No. 50-000-83254