Learn More
Bio Rad Laboratories BIO SAVE
Catalog No. NC9652595
Encompass
Bio-Spin 6 columns contain Bio-Gel P-6 gel in Tris buffer and are used for rapid cleanup of nucleic acid and protein samples. Sample volume is 50-100 microliters. Each disposable polypropylene column contains 1.1 ml of gel.
Catalog No. NC9652595 Supplier Bio Rad Laboratories Supplier No. 1610786
Bio-Spin 6 columns contain Bio-Gel P-6 gel in Tris buffer and are used for rapid cleanup of nucleic acid and protein samples. Sample volume is 50-100 microliters. Each disposable polypropylene column contains 1.1 ml of gel.