Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Current Technologies Bio-Screen™ Biohazard Wipes, Liners, and Rolls

Catalog No. 1441257
Change view
Click to view available options
Dimensions (L x W):
100 ft. x 16 in.
16 x 11 in. (406.4 x 279.4mm)
16 x 16 in. (406.4 x 406.4mm)
3 x 3 in. (76.2 x 76.2mm)
4 x 4 in. (101.6 x 101.6mm)
9 x 8 in. (228.6 x 203.2mm)
Absorbancy:
High
Standard
Super
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Absorbancy
14-412-57 16 x 11 in. (406.4 x 279.4mm) High
14-512-52H 3 x 3 in. (76.2 x 76.2mm) Super
14-412-52D 4 x 4 in. (101.6 x 101.6mm) High
14-412-52C 3 x 3 in. (76.2 x 76.2mm) Standard
14-412-52E 4 x 4 in. (101.6 x 101.6mm) Standard
14-412-4 100 ft. x 16 in. Super
14-412-3 9 x 8 in. (228.6 x 203.2mm) Super
14-412-52B 100 ft. x 16 in. Standard
14-412-58 16 x 16 in. (406.4 x 406.4mm) High
14-412-52G 4 x 4 in. (101.6 x 101.6mm) Super
14-412-52A 100 ft. x 16 in. High
06-670 3 x 3 in. (76.2 x 76.2mm) High
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 14-412-57 Supplier Current Technologies Supplier No. BH11016
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Low-linting, white, absorbent wipes bonded to an orange plastic backing

  • White absorbent side of this wipe / liner is low-linting and more absorbent than gauze or paper wipes
  • Orange plastic backing of wipe / liner blocks soak through of blood, body fluids, specimens and chemicals (xylene, ketones, formalin) to gloves and surfaces
  • Used for cleaning up small / large spills, to wipe pipets, probes, slide prep devices, and to pop caps from blood and specimen tubes
  • Limit cross-contamination from gloves as employees touch phones, keyboards, doors, etc.
  • Larger sizes can be used as mats and liners. Rolls of material can be cut to desired sizes
  • Three thicknesses available: Standard (absorbs 9 times its weight), Heavy (absorbs 12 times its weight), Super (absorbs 25 times its weight)

Specifications

Product Type Biohazard Wipe
Absorbancy High
Length (English) 16 in.
Length (Metric) 406.4 mm
Width (English) 11 in.
Width (Metric) 279.4 mm
Dimensions (L x W) 16 x 11 in. (406.4 x 279.4mm)
Color White and Orange
For Use With (Application) Used for cleaning up small or large spills, to wipe pipets, probes, slide prep devices, and to pop caps from blood and specimen tubes. Wipes and Liners are non-sterile and not intended for use on patients.
Quantity Each
WARNING: Cancer - www.P65Warnings.ca.gov

Wipes and Liners are non-sterile and not intended for use on patients.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.