Learn More
Bio-Serv™ Bottle for Shake and Pour Liquid Diet

Description
- Graduated, reusable plastic bottle with convenient handle, screw-top lid, and wide mouth
- 5 qt. (4.7L) size
Specifications
Specifications
| Capacity (Metric) | 4700 mL |
| Material | Plastic |
| Closure Type | Screw Cap |
| Capacity (English) | 159 oz. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.