Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio-Serv™ Dustless Precision Pellets™ for Primates, Purified
SDP

Catalog No. 14726735
Encompass
Change view
Click to view available options
Quantity:
0.045 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-726-735 0.045 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-726-735 Supplier Bio-Serv™ Supplier No. F0059
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Banana flavored, shown to have superior performance in automatic feeders

  • Precise size and weight pellets generally used in nutritional and behavioral studies
  • Every batch tested for hardness, weight and height
  • Nurtritionally complete purified formula
  • Dustless and fracture free
  • Particle shape: pellet
  • Nutritionally assayed
  • Banana flavored

Nutritional and behavioral studies

Specifications

Product Type Animal Diet
Components Casein, Dextrose, Sucrose, Cellulose, Corn Starch, Corm Oil, Corn Syrup, Mineral Mix, Choline Bitartrate, Calcium Silicate, Banana Flavor, Vitamin Mix, Magnesium Stearate, L-Cystine, Ascorbic Acid
Flavor Banana
Format Pellet
For Use With (Application) Nutritional and behavioral studies
Particle Shape Pellet
Quantity 0.045 g
Species Primate
Storage Requirements Cool dry place
Diameter (English) 0.14 in.
Diameter (Metric) 0.35 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.