Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio-Serv™ Dustless Precision Pellets™ for Rodents, Purified
SDP

Catalog No. p-4277302
Encompass
Change view
Click to view available options
Diameter (English):
0.08 in.
Quantity:
0.015 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (English) Quantity
14-726-748 0.08 in. 0.015 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-726-748 Supplier Bio-Serv™ Supplier No. F05684
Only null left
Add to Cart
Add to Cart

This nutritional treat is used in various rodent studies

  • Precise size and weight pellets generally used in nutritional and behavioral studies
  • Dustless and fracture free
  • Superior performance in automatic feeders
  • Nutritionally complete purified formula
  • Every batch tested for hardness, weight and height
  • Particle shape: pellet
  • Nutritionally assayed

Generally used in nutritional and behavioral studies

Specifications

Product Type Rodent Diet
Calories 3.6
Components Sucrose, Dextrose, Casein, Corn Oil, Mineral Mix, Cellulose, Corn Syrup, Calcium Silicate, Vitamin Mix, Magnesium Stearate, Choline Bitartrate, DL-Methionine, L-Cystine, Ascorbic Acid, Vitamin E Acetate, tBHQ
Flavor Custom
For Use With (Application) Used in nutritional and behavioral studies
Particle Shape Pellet
Quantity 0.015 g
Species Rodent
Storage Requirements Cool dry place
Diameter (English) 0.08 in.
Diameter (Metric) 0.2 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.