Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio-Serv™ Fruit Crunchies™ Animal Diet, Certified
SDP

Catalog No. p-4277306
Encompass
Change view
Click to view available options
Quantity:
1000 g
Sterility:
Non-sterile
Sterile
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Sterility
14-726-772 1000 g Non-sterile
14-726-773 1000 g Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-726-772 Supplier Bio-Serv™ Supplier No. F057981
Only null left
Add to Cart
Add to Cart

A nutritional treat with a medley of flavors

  • Nutritionally complete 190mg treat that compliments enrichment programs
  • A medley of apple, grape and orange flavors add a variety to foraging activity
  • Less labor intensive and more sanitary than perishable fruit
  • Excellent reward or foraging treat
  • Certified (Contaminant Screened)
  • Excellent source of vitamins
  • Nutritionally assayed
  • Weight: 19mg
  • Shape: pellet
  • S05798-1 is sterilized by gamma irradiation

Certifications

Contaminant screened

Specifications

Product Type Fruit Crunchies
Calories 0.66
Components Casein, Dextrose, Sucrose, Cellulose, Corn Starch, Corn Oil, Corn Syrup, Mineral Mix, Choline Bitartrate, Vitamin Mix, Natural and Artificial Flavors, Calcium Silicate, Magnesium Stearate, L-Cystine, Ascorbic Acid, Food Dye
Flavor Fruit Crunchies
Format Pellet
Packaging Quantity 1 kg/bag
Particle Shape Pellet
Quantity 1000 g
Sterility Non-sterile
Storage Requirements Cool dry place
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.