Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio-Serv™ Grainola Trail Mix Foraging Mixture
SDP

Catalog No. 14726403
Encompass
Change view
Click to view available options
Quantity:
1130 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-726-403 1130 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-726-403 Supplier Bio-Serv™ Supplier No. F75711
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Specifications

Product Type Animal Treats
Calories 4.22
Components Rolled Oats, Rolled Wheat, Brown Sugar, Canola Oil, Whey Powder, Coconut, Honey, Almond Flavor and Salt
For Use With (Application) Tuff turf foraging board or scatter into bedding
Particle Shape Dry Mix
Quantity 1130 g
Storage Requirements Cool dry place
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.