Learn More
Bio-Serv™ Ketogenic AIN-76A-Modified Diet, High Fat, Paste
SDP
Catalog No. 14726601
Encompass
Change view
Click to view available options
Quantity:
5000 g
Catalog No. Quantity
14-726-601 5000 g
1 options
Catalog No. 14-726-601 Supplier Bio-Serv™ Supplier No. F3666
High fat, low carbohydrate diet in a paste form
  • Ratio of fat to carbohydrate and protein is approximately 6:1
  • Nutritionally assayed
  • Ingredients: Lard, Butter, Corn Oil, Casein, Cellulose, Mineral Mix, Vitamin Mix, Dextrose
Specifications
Product Type Ketogenic Diet
Calories 7.24
Components Lard, Butter, Corn Oil, Casein, Cellulose, Mineral Mix, Vitamin Mix, Dextrose
Particle Shape Paste
Quantity 5000 g