Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio-Serv™ Lieber-DeCarli '82 Rodent Liquid Diet, Shake and Pour
SDP

Catalog No. 14726631
Encompass
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-726-631 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-726-631 Supplier Bio-Serv™ Supplier No. F1697SP
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

No blender needed—just add water, shake, and pour

  • Easy to prepare
  • Offers the versatility of a liquid format, allowing the addition of alcohol, medications, or test compounds
  • Provides a convenient way to measure food intake or study nutritional manipulation
  • Ideal for post-operative recovery
  • Nutritionally assayed
  • Powder
  • Ingredients: Corn Syrup Solids, Casein, Olive Oil, Maltodextrin, Cellulose, Mineral Mix, Corn Oil, Suspending Aids, Safflower Oil, Vitamin Mix, L-Cystine, Choline Bitartrate, DL-Methionine

Specifications

Product Type Rodent Liquid Diet
Components Corn syrup solids, Casein, Olive Oil, Maltodextrin, Cellulose, Mineral Mix, Corn Oil, Suspending aids, Safflower Oil, Vitamin Mix, L-Cystine, Choline Bitartrate, DL-Methionine
Format Liquid
For Use With (Application) Ideal for post operative recovery
Particle Shape Powder
Quantity 4 L
Species Rodent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.