Learn More
Bio-Serv™ Nutra-Gel Diet™ Grain-Based Formula Dry Mix Kit
SDP
Catalog No. 14726608
Encompass
Change view
Click to view available options
Quantity:
Each
Catalog No. Quantity
14-726-608 Each
1 options
Catalog No. 14-726-608 Supplier Bio-Serv™ Supplier No. F4798KIT
Provides the powdered diet and trays to make Nutra-Gel Diet Grain Based Formula
  • Nutritionally complete grain-based, cherry flavored gel diet
  • Provides both food and water
  • Easy to prepare—just add hot water and mix according to directions
  • Each kit makes 4 trays of 16 cubes at 40g/cube of diet
  • Prepare desired quantities of Nutra-Gel Diet as needed
  • Allows incorporation of drugs into the diet for ease of delivery
  • Ideal for chewing and mobility impairments, post operative care, and weanlings
  • Powder is stable for long transit time in delivery
  • Nutritionally assayed
  • Ingredients: Ground Wheat, Ground Corn, Dehulled Soybean Meal, Wheat Germ, Fish Meal, Brewer's Dried Yeast, Corn Gluten Meal, Porcine Animal Fat (preserved with BHA), Soybean Oil, Gelatin, Mineral Mix, Vitamin Mix, Cherry Flavor, DL-Methionine, Choline Chloride, Sorbic Acid
Specifications
Product Type Animal Diet
Calories 3.74
Components Ground Wheat, Ground Corn, Dehulled Soybean Meal, Wheat Germ, Fish Meal, Brewer's Dried Yeast, Corn Gluten Meal, Porcine Animal Fat (preserved with BHA), Soybean Oil, Gelatin, Mineral Mix, Vitamin Mix, Cherry Flavor, DL-Methionine, Choline Chloride, Sorbic Acid
Flavor Cherry
For Use With (Application) Use for chewing and mobility impairments, post op care and weanlings
Particle Shape Dry Mix
Quantity Each