Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio-Serv™ Nutra-Gel Diet™ Grain-Based Formula, Sterile
SDP

Catalog No. 14726529
Encompass
Change view
Click to view available options
Quantity:
Approx. 40 g/Cube; 16 Cubes/Tray
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-726-529 Approx. 40 g/Cube; 16 Cubes/Tray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-726-529 Supplier Bio-Serv™ Supplier No. S4798TRAY
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Convenient, ready-to-serve diet replaces labor-intensive, nutrient-diluted mush diets

  • Nutritionally complete grain-based, cherry flavored gel diet
  • Provides both food and water
  • Ideal for chewing and mobility impairments, recovering and debilitated rodents, weanlings, and transportation
  • Nutritionally assayed
  • Ingredients: Water, Ground Wheat, Ground Corn, Dehulled Soybean Meal, Wheat Germ, Fish Meal, Brewer's Dried Yeast, Corn Gluten Meal, Porcine Animal Fat (preserved with BHA), Soybean Oil, Gelatin, Mineral Mix, Vitamin Mix, Cherry Flavor, DL-Methionine, Choline Chloride, Sorbic Acid

  • Each cell in tray (Mfr. No. S4798-TRAY) is individually sealed to maintain moisture and sterility

Specifications

Product Type Animal Diet
Calories 1.5
Components Water, Ground Wheat, Ground Corn, Dehulled Soybean Meal, Wheat Germ, Fish Meal, Brewer's Dried Yeast, Corn Gluten Meal, Porcine Animal Fat (preserved with BHA), Soybean Oil, Gelatin, Mineral Mix, Vitamin Mix, Cherry Flavor, DL-Methionine, Choline Chloride, Sorbic Acid
Flavor Cherry
For Use With (Application) Use for chewing and mobility impairments, recovering and debilitated Rodents, weanlings and transportation
Particle Shape Gel
Quantity Approx. 40 g/Cube; 16 Cubes/Tray
Species Rodent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.