Learn More
Bio-Serv™ Nutra-Gel Diet™ Purified Formula Dry Mix Kit
SDP
Catalog No. 14726609
Encompass
Change view
Click to view available options
Quantity:
Each
Catalog No. Quantity
14-726-609 Each
1 options
Catalog No. 14-726-609 Supplier Bio-Serv™ Supplier No. F5769KIT
Provides the powdered diet and trays to make Nutra-Gel Diet Purified Formula
  • Nutritionally complete purified bacon-flavored gel diet
  • Provides both food and water
  • Easy to prepare—just add hot water and mix according to directions
  • Each kit makes 4 trays of 16 cubes at 40g/cube of diet
  • Prepare desired quantities of Nutra-Gel Diet as needed
  • Allows incorporation of drugs into the diet for ease of delivery
  • Ideal for chewing and mobility impairments, post operative care, and weanlings
  • Powder is stable for long transit time in delivery
  • Nutritionally assayed
  • Ingredients: Sucrose, Casein, Corn Starch, Soybean Oil, Gelatin, Cellulose, Mineral Mix, Vitamin Mix, Bacon Flavor, L-Cystine, Xanthan Gum, Sorbic Acid, Choline Bitartrate, tBHQ
Specifications
Product Type Animal Diet
Calories 3.9
Components Sucrose, Casein, Corn Starch, Soybean Oil, Gelatin, Cellulose, Mineral Mix, Vitamin Mix, Bacon Flavor, L-Cystine, Xanthan Gum, Sorbic Acid, Choline Bitartrate, tBHQ
Flavor Bacon
For Use With (Application) Use for chewing and mobility impairments, post op care and weanlings
Particle Shape Dry Mix
Quantity Each