Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio-Serv™ SCIDS MD's Breeders Formula Tablet, Sterile
SDP

Catalog No. 14726332
Encompass
Change view
Click to view available options
Quantity:
100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-726-332 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-726-332 Supplier Bio-Serv™ Supplier No. S0443
May include imposed supplier surcharges.
Add to Cart
Add to Cart

An effective method to reduce the hazards or respiratory problems in immunocompromised mice

  • More convenient and minimizes risk of dehydration compared to using medicated water
  • Contains 60mg (0.002 oz.) Sulfamethoxazole and 12mg (0.0004 oz.) Trimethoprim
  • Nutritionally complete grain-based tablets
  • Sterilized by gamma irradiation
  • Bacon flavored
  • Nutritionally assayed
  • Size: 2.5 dia. x .6cm H (1 x 0.25 in.)
  • 100 per bag

Ingredients: Ground Corn, Banana Flakes, Dehulled Soybean Meal, Corn Gluten Meal, Ground Wheat, Corn Gluten Feed, Fish Meal, Dehydrated Alfalfa Meal, Casein, Dried Whey, Sucrose, Fructose, Dextrose, Dried Beet Pulp, Soybean Oil, Porcine Animal Fat (Preserved with BHA), Dried Brewers Yeast, Mineral Mix, Vitamin Mix, Magnesium Stearate, Calcium Silicate, Bacon Flavor, DL-Methionine, Choline Chloride, Sodium Propionate, Ascorbic Acid, Kaolin, Sulfamethoxazole, Trimethoprim

Specifications

Product Type Diet Supplement
Calories 3.39
Components Ground Corn, Banana Flakes, Dehulled Soybean Meal, Corn Gluten Meal, Ground Wheat, Corn Gluten Feed, Fish Meal, Dehydrated Alfalfa Meal, Casein, Dried Whey, Sucrose, Fructose, Dextrose, Dried Beet Pulp, Soybean Oil, Porcine Animal Fat (preserved with BHA)
Flavor Bacon
Particle Shape Tablet
Quantity 100
Species Mouse
Storage Requirements Cool dry place
Diameter (English) 1 in.
Diameter (Metric) 2.54 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.