Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio-Serv™ Transgenic Dough Diet, Sterile
SDP

Catalog No. 14726517
Encompass
Change view
Click to view available options
Quantity:
1000 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-726-517 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-726-517 Supplier Bio-Serv™ Supplier No. S3472
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nutritionally-complete, soft diet with higher protein and fat levels to comply with NRC and Transgenic-users' recommendations

  • Designed for rodents with chewing, dental or mobility impairments
  • Cookie dough consistency replaces labor intensive, nutrient diluted mush diets
  • Ideal for recovering rodents, weanlings or any time a soft diet is needed
  • Rich bacon flavor stimulates appetite
  • Place in feeder cup or on cage floor
  • Sterilized by gamma irradiation
  • Nutritionally assayed

Ingredients: Sucrose, Whole Milk Powder, Casein, Soybean Oil, Cellulose, Corn Syrup, Vitamin Mix, Mineral Mix, Glycerin, Bacon Flavor, DL-Methionine, Choline Bitartrate, Phenylalanine, tBHQ

Specifications

Product Type Transgenic Dough Diet
Calories 3.83
Components Sucrose, Whole Milk powder, Casein, Soybean Oil, Cellulose, Corn Syrup, Vitamin Mix, Mineral Mix, Glycerin, Bacon Flavor, DL-Methionine, Choline Bitartrate, Phenylalanine, tBHQ
Flavor Bacon
For Use With (Application) Ideal for recovering Rodents, weanlings or any time a soft diet is needed
Particle Shape Dough
Quantity 1000 g
Species Rodent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.