Learn More
Bio Rad Laboratories BIO-SIN 30 COLUMNS 25/PK
Catalog No. NC0152103
Encompass
Micro Bio-Spin 6 columns contain Bio-Gel P-6 gel in Tris buffer and are used for rapid cleanup of nucleic acid and protein samples. Sample volume is 10-75 microliters. Each disposable polypropylene column contains 0.7 ml of gel.
Catalog No. NC0152103 Supplier Bio Rad Laboratories Supplier No. 7326231
Micro Bio-Spin 6 columns contain Bio-Gel P-6 gel in Tris buffer and are used for rapid cleanup of nucleic acid and protein samples. Sample volume is 10-75 microliters. Each disposable polypropylene column contains 0.7 ml of gel.