Learn More
Health Care Logistics Inc Biohazard Specimen Transport Bag, 13 x 18 - 100 Per Package
SDP
Supplier:  Health Care Logistics Inc 18729
Encompass
Biohazard Specimen Transport Bag has a press-and-close adhesive seal that offers liquid-tight, secure sample closure and provides a reliable method of infection protection for staff.Clear transfer bag with scored opening is printed with biohazard logo and instructions for use.Includes separate document pocket with flap.Ideal for multiple or larger specimens.Dimensions: 13" x 18".
Catalog No. NC3135832