Learn More
Bio Rad Laboratories BIO-SPIN 6 TRIS COLUMNS
Catalog No. NC9077443
Encompass
Bio-Spin 30 columns contain Bio-Gel P-30 gel in Tris buffer and are used for rapid cleanup of nucleic acid and protein samples. Sample volume is 50-100 microliters. Each disposable polypropylene column contains 1.1 ml of gel.
Catalog No. NC9077443 Supplier Bio Rad Laboratories Supplier No. 7326227
Bio-Spin 30 columns contain Bio-Gel P-30 gel in Tris buffer and are used for rapid cleanup of nucleic acid and protein samples. Sample volume is 50-100 microliters. Each disposable polypropylene column contains 1.1 ml of gel.