Learn More
Bio Rad Laboratories BIO-SPIN DISP COLUMN
Catalog No. NC9558571
Encompass
Extra thick blot paper is cotton fiber paper that is used in the assembly of a gel and membrane sandwich for electrophoretic transfer, or blotting, applications.
Catalog No. NC9558571 Supplier Bio Rad Laboratories Supplier No. 7326008
Extra thick blot paper is cotton fiber paper that is used in the assembly of a gel and membrane sandwich for electrophoretic transfer, or blotting, applications.