Learn More
Zeflon International, Inc Bio-Tape™ Slides, 25/Bx
SDP
Catalog No. NC1770784
Encompass_Preferred
Bio-Tape™ provides a standardized sampling method for the determination of mold, microbial, bioaerosol, and inorganic dust contamination. It provides the ability to quickly take a sample and measure the relative degree of contamination. It is especially useful when sampling valuable or non-transportable materials and is very effective on smooth areas of concern.
Catalog No. NC1770784 Supplier Zeflon International, Inc Supplier No. BT0025
Bio-Tape™ provides a standardized sampling method for the determination of mold, microbial, bioaerosol, and inorganic dust contamination. It provides the ability to quickly take a sample and measure the relative degree of contamination. It is especially useful when sampling valuable or non-transportable materials and is very effective on smooth areas of concern.