Learn More
Medchemexpress LLC Bio-THZ1 | 1604811-14-4 | 98.1% | 10 MG
SDP
Catalog No. 5000340638
Encompass_Preferred
bio-THZ1 is a biotinylated version of THZ1 that irreversibly binds to CDK7. THZ1 is identified as a selective and potent covalent CDK7 inhibitor with an IC50 of 3.2 nM.
  • Biotinylated version of THZ1
  • Irreversibly binds to CDK7
  • Selective and potent covalent CDK7 inhibitor
  • IC50 of 3.2 nM
Catalog No. 50-003-40638 Supplier Medchemexpress LLC Supplier No. HY12886710MG
bio-THZ1 is a biotinylated version of THZ1 that irreversibly binds to CDK7. THZ1 is identified as a selective and potent covalent CDK7 inhibitor with an IC50 of 3.2 nM.
  • Biotinylated version of THZ1
  • Irreversibly binds to CDK7
  • Selective and potent covalent CDK7 inhibitor
  • IC50 of 3.2 nM