Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bio-THZ1 | 1604811-14-4 | MFCD32201112 | 98.1% | 1053.67 | C52H65ClN12O8S | 5 MG
SDP

Catalog No. 5000262278
Encompass_Preferred

bio-THZ1 is a biotinylated derivative of THZ1 that binds irreversibly to CDK7. It functions as a selective, potent covalent CDK7 inhibitor (THZ1 reported IC50 = 3.2 nM) and is supplied for research use only.

  • Biotinylated derivative for affinity-based assays and pull-downs
  • Irreversible covalent inhibitor targeting CDK7
  • High potency (reported IC50 = 3.2 nM)
  • Molecular weight 1053.67; formula C52H65ClN12O8S
  • Purity approximately 98.06%
  • Highly soluble in DMSO (≈120 mg/mL)
  • Powder storage recommended at -20°C; in solvent storage at -80°C for long term

Catalog No. 50-002-62278 Supplier Medchemexpress LLC Supplier No. HY1288675MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

bio-THZ1 is a biotinylated derivative of THZ1 that binds irreversibly to CDK7. It functions as a selective, potent covalent CDK7 inhibitor (THZ1 reported IC50 = 3.2 nM) and is supplied for research use only.

  • Biotinylated derivative for affinity-based assays and pull-downs
  • Irreversible covalent inhibitor targeting CDK7
  • High potency (reported IC50 = 3.2 nM)
  • Molecular weight 1053.67; formula C52H65ClN12O8S
  • Purity approximately 98.06%
  • Highly soluble in DMSO (≈120 mg/mL)
  • Powder storage recommended at -20°C; in solvent storage at -80°C for long term

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.