Learn More
Alkali Scientific Bio Wand Handheld Personal Sanitizer, Cordless Portable UV Light 265 to 280 nm, Lithium Battery Re chargable USB C
SDP
Catalog No. 5000662554
Encompass_Preferred
The Bio•Wand™ uses UV light (265 to 280 nm) to sanitize objects and surfaces in laboratory environments and general workspaces. This portable device offers a standard one minute cleaning cycle or a more intensive three minute cycle depending on the sanitation level required. To operate, press the power/start button and hold the light approximately 1–2 inches from the surface while slowly moving it across the target area. Allow the UV light to remain on each section for about 5–10 seconds to achieve effective exposure. The Bio•Wand™ provides up to 2 hours of operating time on a full charge and includes a USB C cable for convenient charging and portability. Compact and easy to use, it supports routine sanitation tasks on benches, tools, equipment surfaces, and frequently touched lab items.
Catalog No. 50-006-62554 Supplier Alkali Scientific Supplier No. U1001W
The Bio•Wand™ uses UV light (265 to 280 nm) to sanitize objects and surfaces in laboratory environments and general workspaces. This portable device offers a standard one minute cleaning cycle or a more intensive three minute cycle depending on the sanitation level required. To operate, press the power/start button and hold the light approximately 1–2 inches from the surface while slowly moving it across the target area. Allow the UV light to remain on each section for about 5–10 seconds to achieve effective exposure. The Bio•Wand™ provides up to 2 hours of operating time on a full charge and includes a USB C cable for convenient charging and portability. Compact and easy to use, it supports routine sanitation tasks on benches, tools, equipment surfaces, and frequently touched lab items.