Learn More
Research Products International Corp Biohazard Cryogenic Warning Labels 1.125 x 1.125, 250/roll
SDP
Catalog No. 502229605
Encompass
Biohazard Cryogenic Waterproof labels adhere to plastic, cardboard, tubes, glassware, are tear-proof and able to withstand temperatures from -196°C to +110°C. Preprinted, bright red biohazard warning labels feature crisp black printout on a bright red background. Permanent adhesive labels withstand extreme environments including long-term storage in liquid nitrogen, multiple freeze-thaw cycles and boiling water. Specially laminated, durable labels resist abrasion, exposure to harsh chemicals, solvents and are resistant to UV rays and gamma irradiation (up to 50kGy). Supplied 250 labels per roll.
Catalog No. 50-222-9605 Supplier Research Products International Corp Supplier No. 248505
Biohazard Cryogenic Waterproof labels adhere to plastic, cardboard, tubes, glassware, are tear-proof and able to withstand temperatures from -196°C to +110°C. Preprinted, bright red biohazard warning labels feature crisp black printout on a bright red background. Permanent adhesive labels withstand extreme environments including long-term storage in liquid nitrogen, multiple freeze-thaw cycles and boiling water. Specially laminated, durable labels resist abrasion, exposure to harsh chemicals, solvents and are resistant to UV rays and gamma irradiation (up to 50kGy). Supplied 250 labels per roll.