Learn More
Bio X Cell InVivoMAb Anti-Human c-Myc Monoclonal Antibody, Clone 9E10, 50 MG
Catalog No. NC3861705
Encompass_Preferred
InVivoMAb anti-human c-myc is a purified mouse monoclonal antibody (clone 9E10) that recognizes the human c-Myc epitope tag commonly used for detection and purification of recombinant proteins. It is formulated in PBS without stabilizers and is supplied in multiple sizes, including a 50 mg option for bulk and large-scale laboratory use.
  • Recognizes the c-Myc epitope tag in recombinant proteins.
  • Validated for western blot, ELISA, immunoprecipitation, and flow cytometry.
  • Purified antibody formulated in PBS without stabilizers.
  • Suitable for bulk applications requiring larger quantities.
  • Store at 4C; do not freeze.
Catalog No. NC3861705 Supplier Bio X Cell Supplier No. BE0238R050MG
InVivoMAb anti-human c-myc is a purified mouse monoclonal antibody (clone 9E10) that recognizes the human c-Myc epitope tag commonly used for detection and purification of recombinant proteins. It is formulated in PBS without stabilizers and is supplied in multiple sizes, including a 50 mg option for bulk and large-scale laboratory use.
  • Recognizes the c-Myc epitope tag in recombinant proteins.
  • Validated for western blot, ELISA, immunoprecipitation, and flow cytometry.
  • Purified antibody formulated in PBS without stabilizers.
  • Suitable for bulk applications requiring larger quantities.
  • Store at 4C; do not freeze.