Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio X Cell RecombiMAb Anti-Mouse PD-L1, Clone 10F.9G2-CP154, 1 MG

Catalog No. NC3858827
Encompass_Preferred

The 10F.9G2™-CP154 monoclonal antibody is a recombinant chimeric version of the original 10F.9G2™ antibody, with variable domain sequences identical to the original but constant region sequences switched from rat IgG2b to mouse IgG2a. This antibody contains a LALA-PG mutation in the Fc fragment, preventing it from binding to endogenous Fcγ receptors. It reacts with mouse PD-L1 (programmed death ligand 1), also known as B7-H1 or CD274, and blocks its interaction with PD-1 and B7-1 (CD80).

  • Recombinant chimeric antibody
  • LALA-PG mutation in Fc fragment prevents binding to endogenous Fcγ receptors
  • Blocks interaction between PD-L1 and PD-1, and between PD-L1 and B7-1 (CD80)
  • Purified from CHO cell supernatant in an animal-free facility
  • Purity ≥95%, determined by SDS-PAGE
  • Endotoxin ≤0.5 EU/mg

Catalog No. NC3858827 Supplier Bio X Cell Supplier No. CP154R001MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The 10F.9G2™-CP154 monoclonal antibody is a recombinant chimeric version of the original 10F.9G2™ antibody, with variable domain sequences identical to the original but constant region sequences switched from rat IgG2b to mouse IgG2a. This antibody contains a LALA-PG mutation in the Fc fragment, preventing it from binding to endogenous Fcγ receptors. It reacts with mouse PD-L1 (programmed death ligand 1), also known as B7-H1 or CD274, and blocks its interaction with PD-1 and B7-1 (CD80).

  • Recombinant chimeric antibody
  • LALA-PG mutation in Fc fragment prevents binding to endogenous Fcγ receptors
  • Blocks interaction between PD-L1 and PD-1, and between PD-L1 and B7-1 (CD80)
  • Purified from CHO cell supernatant in an animal-free facility
  • Purity ≥95%, determined by SDS-PAGE
  • Endotoxin ≤0.5 EU/mg

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.