Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio X Cell RecombiMAb anti-mouse VEGF-A, Clone 2G11-2A05-CP061, 1 MG

Catalog No. NC3858829
Encompass_Preferred

The 2G11-2A05-CP061 monoclonal antibody is a recombinant, chimeric antibody with variable domain sequences identical to the original 2G11-2A05. Its constant region was switched from Rat IgG2a, κ to mouse IgG2a, κ for murine models. This antibody reacts with and neutralizes mouse VEGF-A, a 45 kDa homodimeric glycoprotein crucial for angiogenesis and endothelial cell growth.

  • Recombinant, chimeric antibody for reliable and reproducible results
  • Exhibits regulated effector functions and reduced immunogenicity
  • Highly controlled sequence and lack of genetic drift
  • Purity ≥95% (SDS-PAGE); Endotoxin ≤0.5 EU/mg
  • Molecular weight 150 kDa; Sterility by 0.2 μm filtration
  • Purified from mammalian cell supernatant in an animal-free facility

Catalog No. NC3858829 Supplier Bio X Cell Supplier No. CP061R001MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The 2G11-2A05-CP061 monoclonal antibody is a recombinant, chimeric antibody with variable domain sequences identical to the original 2G11-2A05. Its constant region was switched from Rat IgG2a, κ to mouse IgG2a, κ for murine models. This antibody reacts with and neutralizes mouse VEGF-A, a 45 kDa homodimeric glycoprotein crucial for angiogenesis and endothelial cell growth.

  • Recombinant, chimeric antibody for reliable and reproducible results
  • Exhibits regulated effector functions and reduced immunogenicity
  • Highly controlled sequence and lack of genetic drift
  • Purity ≥95% (SDS-PAGE); Endotoxin ≤0.5 EU/mg
  • Molecular weight 150 kDa; Sterility by 0.2 μm filtration
  • Purified from mammalian cell supernatant in an animal-free facility

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.