Learn More
Medchemexpress LLC BIO5192 hydrate 5mg | 5MG
SDP
Supplier:  Medchemexpress LLC HY107589A5MG
Encompass_Preferred
BIO5192 hydrate is a selective and potent integrin 4 1 (VLA-4) inhibitor (Kd 10 pM) BIO5192 hydrate selectively binds to 4 1 (IC50 1 8 nM) over a range of other integrins BIO5192 hydrate results in a 30-fold increase in mobilization of murine hematopoietic stem and progenitors (HSPCs) over basal levels[1 [2
Catalog No. 50-002-41066